ROTO-ROCK GENIE Rotator Rockers
Leverancier: SCIENTIFIC INDUSTRIES ELMIS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
SI-R102
SCIISI-R102EA 2280 EUR
SCIISI-R102
ROTO-ROCK GENIE Rotator Rockers
Schudapparaten Roterende/schommelende schudapparaten
Programmable combination Rotator/Rocker with digital touch screen, featuring magnetic platform and clip plate system.
  • Intuitive touch screen control provides ease when changing program settings
  • Setting the tilt angle and speed control provides thorough mixing while eliminating spills when using open containers in rocking mode
  • Two-sided magnetic platform holds virtually any type of tube or sample vessel when using GENIE clip plates (clip plates sold separately)
  • Programming modes include rock, rotate, or timed auto-switching between the two
  • Compact design
More Product Information