Precision laboratory straight stem thermometers
Leverancier: Ludwig Schneider GmbH & CO. KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1072707 1072808 1082211 1082210 1082209 1082208 1072812 1072811 1072809 1082214 1072807 1072712 1072711 1072710 1072709 1072708 1072810 1082814 1082212 1082213 1082813 1082812 1082811 1082810 1082809 1082808 1052704/03
SCNL1072707EA 58.7 EUR
SCNL1072707 SCNL1072808 SCNL1082211 SCNL1082210 SCNL1082209 SCNL1082208 SCNL1072812 SCNL1072811 SCNL1072809 SCNL1082214 SCNL1072807 SCNL1072712 SCNL1072711 SCNL1072710 SCNL1072709 SCNL1072708 SCNL1072810 SCNL1082814 SCNL1082212 SCNL1082213 SCNL1082813 SCNL1082812 SCNL1082811 SCNL1082810 SCNL1082809 SCNL1082808 SCNL1052704/03
Precision laboratory straight stem thermometers
Thermometers Analoge thermometers
These precision laboratory straight stem thermometers are suitable for official certification. Blue or red filling.
  • Total immersion
  • Colourless prismatic capillary form
  • Upper part Ø: 11 ±0,5 mm; Lower part Ø: 8 ±0,5 mm
Certificaten: Conform to DIN 12781.
Bestelinformatie: Available with works certificate or with DAkkS calibration certificate.