ASTM precision thermometers, ACCU-SAFE
Leverancier: Ludwig Schneider GmbH & CO. KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1205018S 1205012S 1202120S 1202052S 1202049S 1202036S 1202018S
SCNL1205018SEA 146 EUR
SCNL1205018S SCNL1205012S SCNL1202120S SCNL1202052S SCNL1202049S SCNL1202036S 620-2492
ASTM precision thermometers, ACCU-SAFE
Thermometers Analoge thermometers
Thermometer accu for mineral oil and fuel testing.
  • Capillaries form: white back round
  • Ø 6 to 8 mm
  • Liquid Filling: blue
  • Made in Germany
  • Suitable for calibration
Stem type.
Certificaten: Conform to ASTM standards.
Bestelinformatie: Available with works certificate or with DAkkS calibration certificate (optional).
Verpakking: Packed individualy in a square plastic sleeve.