ASTM-Like precision thermometers, ACCU-SAFE
Leverancier: Ludwig Schneider GmbH & CO. KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1202046S 1202119S 1202013S 1202023S 1202024S 1202025S 1202026S 1202027S 1202028S 1202029S 1202041S 1202044S 1202047S 1202048S 1202071S 1202072S 1202073S 1202093S 1202094S 1202095S 1202096S 1202102S 1202103S 1202110S 1202112S 1202118S 1202121S 1202122S 1202123S 1202124S 1202125S 1202126S 1202128S 1202129S 1202132S 1202133S 1205028S
SCNL1202046SEA 244 EUR
SCNL1202046S SCNL1202119S SCNL1202013S SCNL1202023S SCNL1202024S SCNL1202025S SCNL1202026S SCNL1202027S SCNL1202028S SCNL1202029S SCNL1202041S SCNL1202044S SCNL1202047S SCNL1202048S SCNL1202071S SCNL1202072S SCNL1202073S SCNL1202093S SCNL1202094S SCNL1202095S SCNL1202096S SCNL1202102S SCNL1202103S SCNL1202110S SCNL1202112S SCNL1202118S SCNL1202121S SCNL1202122S SCNL1202123S SCNL1202124S SCNL1202125S SCNL1202126S SCNL1202128S SCNL1202129S SCNL1202132S SCNL1202133S SCNL1205028S
ASTM-Like precision thermometers, ACCU-SAFE
Thermometers Analoge thermometers
These precision thermometers are suitable for calibration.
  • Round, white back capillary form
  • Suitable for calibration
Blue filling, stem type.
Leveringsinformatie: With individual serial number.
Verpakking: Each item packed in square plastic sleeve.