ASTM precision thermometers, ACCU-SAFE
Leverancier: Ludwig Schneider GmbH & CO. KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1205088S 1205113S 1205099S 1205016S 1205015S 1205005S 1202117S 1202116S 1202113S 1205014S 1205061S 1205056S 1205054S 1205034S 1205033S 1202099S 1202056S 1202054S 1202061S 1202034S 1202033S 1202088S 1202016S 1202015S 1202014S 1202005S
SCNL1205088SEA 128 EUR
SCNL1205088S SCNL1205113S SCNL1205099S SCNL1205016S SCNL1205015S SCNL1205005S SCNL1202117S SCNL1202116S SCNL1202113S SCNL1205014S SCNL1205061S SCNL1205056S SCNL1205054S SCNL1205034S SCNL1205033S SCNL1202099S SCNL1202056S SCNL1202054S SCNL1202061S 620-2493 620-2461 SCNL1202088S SCNL1202016S 620-2460 SCNL1202014S 620-2457
ASTM precision thermometers, ACCU-SAFE
Thermometers Analoge thermometers
Precision thermometers.
  • Round, white back capillary form
  • Suitable for calibration
Blue filling, stem type.
Certificaten: Conform to ASTM, for mineral oil and fuel testing.
Bestelinformatie: Available with works certificate or with DAkkS calibration certificate (optional).
Verpakking: Packed individualy in a square plastic sleeve.