Precision laboratory thermometers, stem type, IP, ACCU-SAFE
Leverancier: Ludwig Schneider GmbH & CO. KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1212042S
SCNL1212042SEA 147 EUR
SCNL1212042S
Precision laboratory thermometers, stem type, IP, ACCU-SAFE
Thermometers Analoge thermometers
This IP precision thermometer is ideal to measure precisely and reliably without mercury from the very start.
  • Suitable for calibration
  • Blue filling, stem type
Leveringsinformatie: With individual serial number and certificate of conformity.
Verpakking: Each item packed in square plastic sleeve.
Order Now
Specificaties
Environmentally Preferable