Precision ground-joint thermometers
Leverancier: Ludwig Schneider GmbH & CO. KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1292201 1292301 1292202 1292302 1292203 1292303 1292204 1292304 1302210 1292616 1292615 1302212 1302414 1302413 1302412 1302411 1302410 1302216 1302215 1302213 1292611 1302211 1292614 1292613 1292612 1302214 1302415 1302416
SCNL1292201EA 98.7 EUR
SCNL1292201 SCNL1292301 SCNL1292202 SCNL1292302 SCNL1292203 SCNL1292303 SCNL1292204 SCNL1292304 SCNL1302210 SCNL1292616 SCNL1292615 SCNL1302212 SCNL1302414 SCNL1302413 SCNL1302412 SCNL1302411 SCNL1302410 SCNL1302216 SCNL1302215 SCNL1302213 SCNL1292611 SCNL1302211 SCNL1292614 SCNL1292613 SCNL1292612 SCNL1302214 SCNL1302415 SCNL1302416
Precision ground-joint thermometers
Thermometers Analoge thermometers
These precision ground-joint thermometers are suitable for official certification.
  • Colourless, prism capillary form
  • Upper part: Ø 11 ±0,5 mm, with ground joint NS 14,5/23
Blue, red, pentane, alcohol, mercury or toluene filling, enclosed type.
Conforms to DIN 12784.
Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclusively intended to be used for tests as per standards which require the usage of mercury thermometers.
Certificaten: Conforms to DIN 12784.
Bestelinformatie: Available with works certificateMore Product Information