API precision hydrometers
Leverancier: Ludwig Schneider GmbH & CO. KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
3122037S 3122036S 3122040S 3122038S 3122039S 3122045S 3122044S 3122043S 3122042S 3122041S
SCNL3122037SEA 104 EUR
SCNL3122037S SCNL3122036S SCNL3122040S SCNL3122038S SCNL3122039S SCNL3122045S SCNL3122044S SCNL3122043S SCNL3122042S SCNL3122041S
API precision hydrometers
Hydrometers
API precision hydrometers with blue filling thermometer.
Reference temperature: 15,5 °C
Error limit: ±0,1 °API
Certificaten: ASTM E2995-14.
Bestelinformatie: A works or DAkkS calibration certificate is available (on request).
Verpakking: Each packed in a square plastic sleeve.