Precision hydrometers, ASTM E100
Leverancier: Ludwig Schneider GmbH & CO. KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
3222059 3222058 3222057 3222054 3222011 3222007 3222006 3222005 3222004 3222003 3222056 3222001 3222014 3222002 3222021 3222053 3222052 3222051 3222027 3222026 3222025 3222024 3222012 3222022 3222013 3222020 3222019 3222018 3222017 3222016 3222015 3222023 3222055
SCNL3222059EA 49.3 EUR
SCNL3222059 SCNL3222058 SCNL3222057 SCNL3222054 SCNL3222011 SCNL3222007 SCNL3222006 SCNL3222005 SCNL3222004 SCNL3222003 SCNL3222056 SCNL3222001 SCNL3222014 SCNL3222002 SCNL3222021 SCNL3222053 SCNL3222052 SCNL3222051 SCNL3222027 SCNL3222026 SCNL3222025 SCNL3222024 SCNL3222012 SCNL3222022 SCNL3222013 SCNL3222020 SCNL3222019 SCNL3222018 SCNL3222017 SCNL3222016 SCNL3222015 SCNL3222023 SCNL3222055
Precision hydrometers, ASTM E100
Hydrometers
These precision ASTM E100 hydrometers are designed for specific gravity or relative density.
  • Reference temperature: 15,5 °C (60/60 °F)
  • Without thermometer
  • Suitable for calibration
Bestelinformatie: Available with either a works or a DAkkS calibration certificate on request
Verpakking: Each packed in a square plastic sleeve.