Precision thermometers
Leverancier: Ludwig Schneider GmbH & CO. KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
64346 64345 64341 64340 64335 64336
SCNL64346EA 56.8 EUR
SCNL64346 SCNL64345 SCNL64341 SCNL64340 SCNL64335 SCNL64336
Precision thermometers
Thermometers Analoge thermometers
Precision thermometer, round, white back capillary form, red filling, stem type.
  • With permanent graduation and lettering
  • With serial number and certificate (conformance and accuracy)
Verpakking: Single packed in plastic square shucks.