Exact-Temp thermometers
Leverancier: Ludwig Schneider GmbH & CO. KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
8000111 8000112 8000113 8000114 8000115 8010111 8010112 8010113 8010114 8010115 8010116 7010112 8000117 8000116 7000117 7010111 7010116 7010115 7010114 7010113 7000114 7000115 7000113 7000112 7000111 7000116 7000111/04/2P 7010111/04/2P 7000112/04/2P 7010112/04/2P 7000114/04/2P 7000113/04/2P 7010113/04/2P 7010114/04/2P 7000115/04/2P 7010115/04/2P 7000116/04/2P 7010116/04/2P 8010112/04/2P
SCNL8000111EA 74.8 EUR
SCNL8000111 SCNL8000112 SCNL8000113 SCNL8000114 SCNL8000115 SCNL8010111 SCNL8010112 SCNL8010113 SCNL8010114 SCNL8010115 SCNL8010116 SCNL7010112 SCNL8000117 SCNL8000116 SCNL7000117 SCNL7010111 SCNL7010116 SCNL7010115 SCNL7010114 SCNL7010113 SCNL7000114 SCNL7000115 SCNL7000113 SCNL7000112 SCNL7000111 SCNL7000116 SCNL7000111/04/2P SCNL7010111/04/2P SCNL7000112/04/2P SCNL7010112/04/2P SCNL7000114/04/2P SCNL7000113/04/2P SCNL7010113/04/2P SCNL7010114/04/2P SCNL7000115/04/2P SCNL7010115/04/2P SCNL7000116/04/2P SCNL7010116/04/2P SCNL8010112/04/2P
Exact-Temp thermometers
Thermometers Analoge thermometers
Red or blue filling. Coating: PTFE/FEP, transparent.
  • Available as digital or conventional thermometers (organic or mercury filling, with/without safety coating)
  • Unbreakable plastic bottles, attached magnets for easy wall or door mounting, individual serial number
  • All thermometers are encapsulated in a high grade safety coating to protect contents, users and environment from splintered glass or display fluid leakage (PTFE/FEP, transparent)
High precision instruments to monitor freezers, ultra-low freezers, blood bank freezers, refrigerators, incubatorsMore Product Information