UV ring lights, VisiLED, S80-55UV
Leverancier: SCHOTT AG Lighting and imaging
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
400255 400225 400200 400266
630-3504EA 879 EUR
630-3504 SCOC400225 630-3180 SCOC400266
UV ring lights, VisiLED, S80-55UV
Lampen Ringlichten
The unique VisiLED UV ring light allows electronic switching between bright field and UV.
  • Segment ring light, consists of 4 UV and 4 white LED segments
  • The only segment ring light in which white-light and UV-LEDs are alternately installed in eight segments
  • LED arrangement allows objects to be examined from the same illumination angle, significantly improving the ability to compare and reproduce resulting images
  • While the homogenous white light displays samples in natural color, UV excitation reveals previously invisible parts of the sample through