Verdunnings viscositeitsmeters
Leverancier: SI Analytics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
285401403 285401411 285401428 285401444 285401452
SCOI285401403EA 431 EUR
SCOI285401403 616-0031 SCOI285401428 SCOI285401444 SCOI285401452
Verdunnings viscositeitsmeters
Viscosimeters Capillaire viscometers
Ubbelohde viscometers with suspended ball level used to determine the limit viscosity of polymers.
The limit number is determined in combination with the viscosity measuring instruments and piston burette.