Glass tubings, DURAN® borosilicate glass
Leverancier: Schott AG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1026643 1026165 1026159 1026072 1026020 1026221 1026642 1026190 1026186 1026201 1026171 1026645 1026193 1026630 1026237 1026210 1026226 1026183 1026160 1026953 1026955 1026224 1026080 1026961 1026176 1026634 1026055 1026195 1026203 1026236 1026189 1026635 1026234 1026231 1026200 1026959 1026185 1026188 1026957 1026958 1026240 1026192 1026181 1026161 1026233 1026162 1026638 1026644 1026208 1026213 1026225 1026239 1026956 1026654 1026196 1026222 1026229 1026260 1026960 1026223 1026230 1026629 1026227 1026243 1026187 1026163 1026954 1026172 1026169 1026632 1026168 1026235 1026215 1026242 1026180 1026241 1026952 1026218 1026199 1026217 1026220 1026238 1026636 1026207 1026143 1026179 1026197 1026216 1026178 1026228 1185645 1114929 1026349
SCOR1026643EA 695 EUR
SCOR1026643 SCOR1026165 SCOR1026159 SCOR1026072 SCOR1026020 SCOR1026221 SCOR1026642 SCOR1026190 SCOR1026186 SCOR1026201 SCOR1026171 SCOR1026645 SCOR1026193 SCOR1026630 SCOR1026237 SCOR1026210 SCOR1026226 SCOR1026183 SCOR1026160 SCOR1026953 SCOR1026955 SCOR1026224 SCOR1026080 SCOR1026961 SCOR1026176 SCOR1026634 SCOR1026055 SCOR1026195 SCOR1026203 SCOR1026236 SCOR1026189 SCOR1026635 SCOR1026234 SCOR1026231 SCOR1026200 SCOR1026959 SCOR1026185 SCOR1026188 SCOR1026957 SCOR1026958 SCOR1026240 SCOR1026192 SCOR1026181 SCOR1026161 SCOR1026233 SCOR1026162 SCOR1026638 SCOR1026644 SCOR1026208 SCOR1026213 SCOR1026225 SCOR1026239 SCOR1026956 SCOR1026654 SCOR1026196 SCOR1026222 SCOR1026229 SCOR1026260 SCOR1026960 SCOR1026223 SCOR1026230 SCOR1026629 SCOR1026227 SCOR1026243 SCOR1026187 SCOR1026163 SCOR1026954 SCOR1026172 SCOR1026169 SCOR1026632 SCOR1026168 SCOR1026235 SCOR1026215 SCOR1026242 SCOR1026180 SCOR1026241 SCOR1026952 SCOR1026218 SCOR1026199 SCOR1026217 SCOR1026220 SCOR1026238 SCOR1026636 SCOR1026207 SCOR1026143 SCOR1026179 SCOR1026197 SCOR1026216 SCOR1026178 SCOR1026228 SCOR1185645 SCOR1114929 SCOR1026349
Glass tubings, DURAN® borosilicate glass
Slangen Transfer Tubing
Borosilicate glass acc. to ISO 3585, all-purpose glass for apparatus for the chemical industry, for pipelines and laboratory.
  • High transmittance
  • Electrically highly insulating
  • High chemical resistance
  • High hydrolytic resistance
  • High resistance to thermal shock
  • High temperature resistance
Tube length ~1500 mm.