Glass tubings, DURAN® borosilicate glass
Leverancier: Schott AG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1026996 1026245 1026089 1026972 1026108 1026247 1026103 1026258 1026969 1026872 1026964 1026120 1026963 1026965 1026244 1026993 1026871 1026979 1026267 1026985 1026115 1026980 1026255 1026253 1026998 1026977 1026967 1026966 1026264 1026989 1026252 1026974 1026248 1026870 1026251 1026249 1026970 1026109 1026988 1026114 1026968 1026257 1026262 1026984 1026987 1026990 1026256 1026263 1026111 1026994 1026119 1026973 1026978 1026268 1026975 1026084 1026266 1026246 1026992 1026254 1026995 1026265 1026983 1026991 1025260 1026261 1026672 1026962 1026971 1026976 1026981 1026982 1026986 1026085 1026091 1026096 1026107
SCOR1026996EA 1110 EUR
SCOR1026996 SCOR1026245 SCOR1026089 SCOR1026972 SCOR1026108 SCOR1026247 SCOR1026103 SCOR1026258 SCOR1026969 SCOR1026872 SCOR1026964 SCOR1026120 SCOR1026963 SCOR1026965 SCOR1026244 SCOR1026993 SCOR1026871 SCOR1026979 SCOR1026267 SCOR1026985 SCOR1026115 SCOR1026980 SCOR1026255 SCOR1026253 SCOR1026998 SCOR1026977 SCOR1026967 SCOR1026966 SCOR1026264 SCOR1026989 SCOR1026252 SCOR1026974 SCOR1026248 SCOR1026870 SCOR1026251 SCOR1026249 SCOR1026970 SCOR1026109 SCOR1026988 SCOR1026114 SCOR1026968 SCOR1026257 SCOR1026262 SCOR1026984 SCOR1026987 SCOR1026990 SCOR1026256 SCOR1026263 SCOR1026111 SCOR1026994 SCOR1026119 SCOR1026973 SCOR1026978 SCOR1026268 SCOR1026975 SCOR1026084 SCOR1026266 SCOR1026246 SCOR1026992 SCOR1026254 SCOR1026995 SCOR1026265 SCOR1026983 SCOR1026991 SCOR1025260 SCOR1026261 SCOR1026672 SCOR1026962 SCOR1026971 SCOR1026976 SCOR1026981 SCOR1026982 SCOR1026986 SCOR1026085 SCOR1026091 SCOR1026096 SCOR1026107
Glass tubings, DURAN® borosilicate glass
Slangen Transfer Tubing
Borosilicate glass acc. to ISO 3585, all-purpose glass for apparatus for the chemical industry, for pipelines and laboratory.
  • High transmittance
  • Electrically highly insulating
  • High chemical resistance
  • High hydrolytic resistance
  • High resistance to thermal shock
  • High temperature resistance
Tube length ~1500 mm.
Order Now
Specificaties
Environmentally Preferable