Flessen, smalle hals met genormaliseerde slijping, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
211647301 211648606 211649105 211643608 211642406 211644407 211645403 211646305 211641701
SCOT211647301EA 215 EUR
SCOT211647301 SCOT211648606 SCOT211649105 SCOT211643608 SCOT211642406 SCOT211644407 SCOT211645403 SCOT211646305 SCOT211641701
Flessen, smalle hals met genormaliseerde slijping, DURAN®
Flessen Glazen flessen
DURAN®, borosilicate glass 3.3, clear or brown, with standard ground joint, without stopper.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Glass Type I according to USP, EP and JP
Brown tinted to protect light-sensitive substances.
With printed retrace code (batch identification).