Cylinders, borosilicate glass, with hexagonal base
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
213910776 213910879 213911472 213911772 213912477 213913679 213914478 213915474 213916376
SCOT213910776EA 81.7 EUR
SCOT213910776 SCOT213910879 SCOT213911472 SCOT213911772 SCOT213912477 SCOT213913679 SCOT213914478 SCOT213915474 SCOT213916376
Cylinders, borosilicate glass, with hexagonal base
Cilinders Maatcilinders
DURAN® borosilicate glass 3.3, clear.
  • Without graduation
  • Hexagonal base
  • Base with three knobs which increase its stability
Certificaten: DIN EN ISO 4788.
Order Now
Specificaties
Environmentally Preferable