DURAN® Measuring Cylinders
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
213970708 213970802 213971404 213971704 213972409 213973602 213974401 213975406 213976308
SCOT213970708EA 155 EUR
SCOT213970708 SCOT213970802 SCOT213971404 SCOT213971704 SCOT213972409 SCOT213973602 SCOT213974401 SCOT213975406 SCOT213976308
DURAN® Measuring Cylinders
Cilinders Maatcilinders
Class A, delivered with UKAS certificate.
  • Made from borosilicate 3.3 glass
  • Individually tested and supplied with a calibration certificate complying to the requirements of ISO 17025 and is accredited the United Kingdom Accreditation Service (UKAS)
  • The volume content tolerances conform to accuracy Class A
  • Easy-to-read scale with graduation marks to estimate the content
  • The blue printed graduations are highly chemically resistant
DURAN® volumetric measuring instruments already meet the highest accuracy standard. The DURAN® range has now been expanded toMore Product Information
Order Now
Specificaties
Environmentally Preferable