Mixing Measuring Cylinders, Tall Form
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
214140874 214141476 214141776 214142472 214143674 214144473 214145478 214146371
SCOT214140874EA 112 EUR
SCOT214140874 SCOT214141476 SCOT214141776 SCOT214142472 SCOT214143674 SCOT214144473 SCOT214145478 SCOT214146371
Mixing Measuring Cylinders, Tall Form
Cilinders Maatcilinders
DURAN®, borosilicate glass 3.3, clear.
  • Without graduation
  • Without stopper
  • Very good chemical and temperature resistance
With hexagonal base to prevent the cylinder from rolling and standard ground joint.
Certificaten: DIN EN ISO 4788.