Oil centrifuge tubes, conical, short and long cone, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
216032516
SCOT216032516EA 347 EUR
SCOT216032516
Oil centrifuge tubes, conical, short and long cone, DURAN®
Buisjes Centrifugebuisjes
DURAN® oil centrifuge tubes are used for the determination of water and sediments in crude oils.
  • Markings and graduations are lasered and therefore have excellent resistance to aggressive solvents
  • Legibility and line uniformity across the entire scale range
  • Calibrated to contain
  • The closest volumetric measurement tolerance exists compared to competing products
  • Made of borosilicate glass 3.3
  • Autoclavable
Crude oil (also often called petroleum) is one of the most important raw materials and is used to manufacture a wide range of chemical products suchMore Product Information