Adapter, ontvanger, vacuüm, gebogen, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
241302105 241303401 241304603
SCOT241302105EA 58.4 EUR
SCOT241302105 SCOT241303401 SCOT241304603
Adapter, ontvanger, vacuüm, gebogen, DURAN®
Onderdelen voor destillatie Destillatieadapters
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Bent, with two standard ground joints and screw-on plastic hose connection.