Condensors, Dimroth, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
242546101 242547106 242547209 242548205
SCOT242546101EA 83.6 EUR
SCOT242546101 SCOT242547106 201-3335 SCOT242548205
Condensors, Dimroth, DURAN®
Condensors
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For condensing vapours. Water supplied through plastic connectors.
The Dimroth condenser comprises a coil condenser located within a tube. This condenser type has a larger heat exchange surface and thus a better cooling effect than the Liebig or Allihn condenser.