Droogbuizen met mannelijk slijpstuk, gebogen, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
242620609 242620703 242620806 242620909
SCOT242620609EA 27.9 EUR
SCOT242620609 SCOT242620703 SCOT242620806 SCOT242620909
Droogbuizen met mannelijk slijpstuk, gebogen, DURAN®
Buisjes Droogbuizen
DURAN® borosilicate glass 3.3. clear. Angled at 75° with ground joint cone.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Suitable for use with a desiccant such as calcium chloride.