DURAN® Burettes, Straight Standard Ground Stopcock, Class AS
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
243211109 243211606 243212208 243212705
SCOT243211109EA 447 EUR
SCOT243211109 SCOT243211606 SCOT243212208 SCOT243212705
DURAN® Burettes, Straight Standard Ground Stopcock, Class AS
Buretten
The DURAN® burettes are made of highly chemically resistant borosilicate glass 3.3. Their precise scaling makes them particularly suitable for titrimetric purposes. The amount of liquid required for titration, but previously unknown, can be read off very accurately after the reaction has been completed.
  • Made from borosilicate glass 3.3
  • Graduated, main volumes ring marked
  • Calibration is based on the poured out volume ('Ex') at a 20 °C reference temperature
  • The volume content tolerances conform to accuracy Class AS
  • With Schellbach stripe and straight
More Product Information
Order Now
Specificaties
Environmentally Preferable