Kranen, éénweg, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
286402402 286403407 286403604 286404806 286410805 286411604 286412009 286412403 286412609 286412806 286413408 286413605 286413802 286414601 286414807 286414207 286405408 286406807 286407203 286407409 286407606 286407803 286415006
SCOT286402402EA 215 EUR
SCOT286402402 SCOT286403407 SCOT286403604 SCOT286404806 SCOT286410805 SCOT286411604 SCOT286412009 SCOT286412403 201-0383 SCOT286412806 SCOT286413408 SCOT286413605 SCOT286413802 SCOT286414601 SCOT286414807 SCOT286414207 SCOT286405408 SCOT286406807 SCOT286407203 SCOT286407409 SCOT286407606 SCOT286407803 SCOT286415006
Kranen, éénweg, DURAN®
Afsluitkranen
DURAN®, borosilicate glass 3.3, clear, with glass or PTFE keys.
  • Replaceable NS keys
  • Screw thread retaining device
Straight bore.