Brushes, for NMR tubes, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
290740059
SCOT290740059EA 3.3 EUR
SCOT290740059
Brushes, for NMR tubes, DURAN®
Borstels
Stubborn soiling and deposits in NMR tubes can be easily removed with the NMR brush.
  • Due to the accuracy of fit, bristles rest against the inner wall of the glass for an optimal cleaning process
  • Recommended usable temperature range is –70 to 200 °C
  • Stainless steel wire with nylon bristles, curved end of bristle trim
Nuclear magnetic resonance spectroscopy (NMR-spectroscopy) is a spectroscopic method to elucidate the structure of molecules and compounds. This analytical method requires very precise tubes with a constant diameter, a uniform wallMore Product Information