Grounding mat and cables
Leverancier: S.C.A.T.
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
108009 108011 107938 107939 108099 117982 108096 108092 108275 108222 108268
590-0699EA 39.9 EUR
590-0699 590-0701 590-0637 590-0638 SCTV108099 SCTV117982 SCTV108096 SCTV108092 SCTV108275 SCTV108222 SCTV108268
Grounding mat and cables
Aardingen Aardingskabels
In order to prevent static discharge with fluids in plastic containers, the contents of the waste receptacles can be secured with ground connection. This allows users to establish an additional safety factor when collecting combustible fluids.
  • Grounding cable with different connection types
  • Cable lengths from 1,5 to 3 metres available
  • Matching with Grounding plug EU 1 megohm for connection to building services
With the different grounding cables users have the possibility to safely conduct the electrostatic charge and prevent the risk of explosion dueMore Product Information