Waste disposal-sets, V2.0
Leverancier: S.C.A.T.
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
307416 307315 307320 307321
SCTV307416EA 449 EUR
SCTV307416 SCTV307315 SCTV307320 SCTV307321
Waste disposal-sets, V2.0
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
These safety funnels and containers are used all over the world wherever the safe disposal of liquid waste is important. To ensure that collection points are also quickly ready for use, the most popular products have been assembled as sets. This requires only one part number for the comprehensive safety system, guaranteed compatible without time-consuming assembly of individual components.
  • Set components made of high quality and durable PE-HD material
  • Electrostatic conductive and therefore protected against ignition and explosion
  • Lid funnel with removable
More Product Information