Hand and body wash lotions, Sensiva®
Leverancier: SCHUELKE MAYR
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
114005 114003
SCUE114005EA 0 EUR
SCUE114005 SCUE114003
Hand and body wash lotions, Sensiva®
Huidverzorging Zepen en huidreinigingsmiddelen
Gentle, soap-free wash lotion with skin-compatible surfactant for nourishing hand and body cleaning.
  • Contains Allantoin which has a soothing effect on the skin, moisturises and supports cell renewal
  • Dermatologically tested
  • Hand cleaning before surgical and hygienic hand disinfection
  • Skin-friendly colour, perfume and lanolin-free-free
Ideally suitable for the food-processing industry.
Opgelet: *De product(en) op deze pagina vallen buiten het toepassingsgebied van de EU-richtlijn 2019/904