Bouffant dispenser with lid
Leverancier: S-CURVE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BD-600
SCURBD-600EA 184 EUR
SCURBD-600
Bouffant dispenser with lid
PBM-dispensers
Bouffant dispenser, made of clear acrylic, for cleanroom bouffants.
  • Single compartment
  • Lid with full length piano hinge
  • Large round opening for easy access
  • Flame treated edges for a smooth finish
  • Keyholes for wall mounting