Packaging, bottles, narrow neck, square, HDPE, with screw caps
Leverancier: SEMADENI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2022
SEMA2022EA 603 EUR
SEMA2022
Packaging, bottles, narrow neck, square, HDPE, with screw caps
Flessen Kunststof flessen
HDPE, translucent.
  • Good chemical resistance
  • Good temperature resistance
Bestelinformatie: Screw caps must be ordered separately.