Polyurethane tubings, transparent
Leverancier: SEMADENI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2591
SEMA2591EA 28.8 EUR
SEMA2591
Polyurethane tubings, transparent
Slangen Transfer Tubing
Made of PU.
  • High mechanical strength
  • Resistant to dilute acids and alkaline solutions
Suitable for pressure lines, clean-up and analysis.
Operating temperature range −50 to 80 °C.