Carboys, wide neck, LDPE
Leverancier: SEMADENI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
3060
SEMA3060EA 396 EUR
SEMA3060
Carboys, wide neck, LDPE
Flessen Voorraadflessen
LDPE, translucent.
  • Good chemical resistance
  • Good temperature resistance
With silicone tubing, pinchcock and screw cap.
Order Now
Specificaties
Environmentally Preferable