Carboys, wide neck, with screw caps
Leverancier: SEMADENI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5554
SEMA5554EA 101 EUR
SEMA5554
Carboys, wide neck, with screw caps
Flessen Voorraadflessen
HDPE, translucent.
  • Good chemical resistance
  • Good temperature resistance
With blue screw cap and blue handle.