5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na)

Leverancier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

15259.01 15259.02
SERA15259.01EA 0 EUR
SERA15259.01 SERA15259.02
5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na)
5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na)
Formule: C₈H₄BrClNNa₂O₄P
MDL Nummer: MFCD00036757
CAS nummer: 102185-33-1

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR