Penicillin G potassium salt

Leverancier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

31749.03 31749.04
SERA31749.03EA 85 EUR
SERA31749.03 SERA31749.04
Penicillin G potassium salt
Penicillin G potassium salt
Formule: C₁₆H₁₇KN₂O₄S
Smeltpunt: 214…217 °C
Dichtheid: 1,359 g/cm³ (20 °C)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00036193
CAS nummer: 113-98-4
EINECS: 204-038-0
Merck Index: 12,07225

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR