Polyethylene glycol 40,000

Leverancier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

33139.01 33139.02
SERA33139.01EA 34 EUR
SERA33139.01 SERA33139.02
Polyethylene glycol 40,000
Polyethylene glycol 40,000
Formule: H(OCH₂CH₂)nOH
Bewaar temperatuur: Kamertemperatuur
CAS nummer: 25322-68-3
EINECS: 500-038-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR