Silicone fluid, 50 cSt DC 200

Leverancier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Siliconenolie

35134.01
SERA35134.01EA 47 EUR
SERA35134.01
Silicone fluid, 50 cSt DC 200
Silicone fluid

Silicone DC 200 fluid


Leveringsinformatie: This product is not available in all countries. Please check with your local VWR International office or supplier.

Dichtheid: 0.963 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00084411
CAS nummer: 63148-62-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR