Tetrazolium blue chloride

Leverancier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

35950.02 35950.04
SERA35950.02EA 0 EUR
SERA35950.02 SERA35950.04
Tetrazolium blue chloride
Tetrazolium blue chloride
Formule: C₄₀H₃₂Cl₂N₈O₂
Smeltpunt: 265…270 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00040933
CAS nummer: 1871-22-3
EINECS: 217-488-8
Merck Index: 12,09380

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR