Dispenser and pump syringes
Leverancier: Trajan Scientific and Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
004696 005696 006696 007693 007696 008193 008196 008593 008596 008793 008796 008996 009496
SGEA004696EA 149 EUR
SGEA004696 SGEA005696 SGEA006696 SGEA007693 SGEA007696 SGEA008193 SGEA008196 SGEA008593 SGEA008596 SGEA008793 SGEA008796 SGEA008996 SGEA009496
Dispenser and pump syringes
Spuiten Injectiespuiten voor chromatografie
These dispenser and pump syringes incorporated with Diamond technology have been designed and manufactured for a variety of instruments and customer applications.
  • Improve the accuracy and precision of results
  • Robust and durable for longer life
  • Less maintenance downtime and reduced maintenance costs
  • Precise fit with greater reliability
  • Do not contaminate the aspirated fluid
  • Increases flexibility with a broad functional temperature range and high chemical resistance