Capillary columns, BPX-DXN
Leverancier: Trajan Scientific and Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
054235
SGEA054235EA 1290 EUR
SGEA054235
Capillary columns, BPX-DXN
Kolommen voor chromatografie
A high temperature, low bleed, highly inert GC column for the analysis of dioxins and related compounds.
  • Ideal for trace analysis and GC-MS
BPX-DXN is ideal for applications such as persistent organic pollutants (POPs), polychlorinated biphenyls (PCBs), polychlorinated dibenzofurans (PCDFs) and polychlorinated dibenzo-p-dioxins (PCDDs).