Capillary electrophoresis tubing
Leverancier: Trajan Scientific and Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
062801 062803 062812 062813 062823 062881
SGEA062801EA 53.3 EUR
SGEA062801 SGEA062803 SGEA062812 SGEA062813 SGEA062823 SGEA062881
Capillary electrophoresis tubing
Slangen Chromatography Tubing
Manufactured from precision bore tubing.
  • Hydrophobic and hydrophilic surface treatments
  • Wide pH range
  • 30, 50, 75 and 100 µm Int.Ø
  • Column-to-column reproducibility