Micro control valves
Leverancier: Trajan Scientific and Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1236091 1236103 1236104 1236282 1236283
SGEA1236091EA 681 EUR
SGEA1236091 SGEA1236103 SGEA1236104 SGEA1236282 SGEA1236283
Micro control valves
Ventielen Terugslagventielen
Micro control valves provide precision metering and on-off control of gas flows. They are designed specifically for chromatography applications and used at elevated temperatures from high vacuum to medium pressure.
Actuated remotely with a toggle switch or solenoid valve, and a 50 to 55 psi air supply.