CRS Endura-Seals GC septa
Leverancier: Trajan Scientific and Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
250101 250111
SGEA250101EA 115 EUR
SGEA250101 SGEA250111
CRS Endura-Seals GC septa
Tussenschoten
Endura-Seals are the CRS alternative to typical silicone-rubber injection port septa for gas chromatography. They work in a 2 part sealing system that includes the septa and the sealing nut.
  • Reliably seal against the carrier gas pressure in the inlet
  • Capable of being pierced and resealing time after time
  • Does not contaminate or bleed material into the chromatographic system
  • Resilient to needle wear and tear
  • Eliminates coring
The Endura-Seals are available in 11 mm and Shimadzu style kits. Each kit includes 1 sealing nut and 2 septa.