HyClone™ Standard Fetal Bovine Serum (FBS), U.S. Origin, HyClone products (Cytiva)

Leverancier: HyClone products (Cytiva)
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

SH30088.02 SH30088.03 SH30088.02HI SH30088.03HI SH30088.03IR25
HYCLSH30088.02EA 124 EUR
HYCLSH30088.02 HYCLSH30088.03 HYCLSH30088.02HI HYCLSH30088.03HI SH30088.03IR25
HyClone™ Standard Fetal Bovine Serum (FBS), U.S. Origin, HyClone products (Cytiva)
Serum
HyClone™ standard cell culture grade FBS is filtered through three sequential 100 µm (0,1 µm) pore-size rated filters.

  • Sourced in the USA with complete traceability back to original source
  • Low in antibodies and high in growth factors
  • Produced using true pool technology for uniformity and consistency from bottle to bottle
  • Filtered through three sequential 100 nm (0.1 µm) pore size-rated filters

HyClone Standard Fetal Bovine Serum (FBS), U.S. origin is an economical alternative to characterized and defined FBS. Standard FBS is ideal for researchMore Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR