HyClone™ Fetal Bovine Serum (United States), Super Low IgG, HyClone Products (Cytiva)
Leverancier: HyClone products (Cytiva)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
SH30898.02 SH30898.03 SH30898.02HI SH30898.03HI SH30898.03IH25 SH30898.03IR25 SH30898.01
HYCLSH30898.02EA 181 EUR
HYCLSH30898.02 HYCLSH30898.03 HYCLSH30898.02HI HYCLSH30898.03HI SH30898.03IH25 SH30898.03IR25 SH30898.01
HyClone™ Fetal Bovine Serum (United States), Super Low IgG, HyClone Products (Cytiva)
Serum
Enhance cell culture growth with U.S.-sourced Cytiva HyClone Super Low IgG Fetal Bovine Serum (FBS).
  • Processed via protein G chromatography
  • Enhance cell culture growth
  • U.S. sourced
Enhance cell culture growth with U.S.-sourced Cytiva HyClone Super Low IgG Fetal Bovine Serum (FBS), processed via protein G chromatography to substantially reduce immunoglobulin G (IgG) levels.