Human Recombinant IGF-II (from E. coli)
Leverancier: Shenandoah Biotechnology
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
100-14-100UG 100-14-10UG 100-14-1MG 100-14-500UG 100-14-50UG
SHBT100-14-100UGEA 327 EUR
SHBT100-14-100UG SHBT100-14-10UG SHBT100-14-1MG SHBT100-14-500UG SHBT100-14-50UG
Human Recombinant IGF-II (from E. coli)
Eiwitten en peptiden
Insulin-like growth factor II (IGF-II) is an important fetal growth hormone made by theca cells during gestation. IGF-II engages the IGF-I receptor (IGF1R) to mediate embryonic growth. IGF-II also binds the sink IGF-II receptor (IGF2R) leading to IGF-II degradation.
  • High quality
  • Low endotoxin
  • Affordable
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
Leveringsinformatie: A certificate of analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
Opgelet: For research use only.