Rat Recombinant MIP-1 A (from E. coli)
Leverancier: Shenandoah Biotechnology
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
300-23-100UG 300-23-1MG 300-23-20UG 300-23-500UG 300-23-5UG
SHBT300-23-100UGEA 453 EUR
SHBT300-23-100UG SHBT300-23-1MG SHBT300-23-20UG SHBT300-23-500UG SHBT300-23-5UG
Rat Recombinant MIP-1 A (from E. coli)
Eiwitten en peptiden
Interleukin 17F (IL-17F), a member of the IL-17 cytokine family, is secreted by activated CD4+ T cells and monocytes. IL-17F binds the IL-17 receptor related molecule, IL17RC, to promote the production of the interleukin 6 (IL-6), interleukin 8 (IL-8), and granulocyte macrophage colony-stimulating factor (GM-CSF) cytokines.
  • High quality
  • Low endotoxin
  • Affordable
IL-17F also functions to regulate matrix turnover rates, inhibit endothelial cell angiogenesis, and induce the endothelial cell expression of interleukin 2 (IL-2), monocyte chemoattractant protein-1 (MCP-1), and transforming growth factor beta 1 (TGF-β1).
Leveringsinformatie: A certificate of analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
Opgelet: For research use only.