IP-10 (from E. coli)
Leverancier: Shenandoah Biotechnology
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
400-02-100UG 400-02-1MG 400-02-25UG 400-02-500UG 400-02-5UG
SHBT400-02-100UGEA 453 EUR
SHBT400-02-100UG SHBT400-02-1MG SHBT400-02-25UG SHBT400-02-500UG SHBT400-02-5UG
IP-10 (from E. coli)
Eiwitten en peptiden
US-made bioactive recombinant proteins offer rigid quality testing and low manufacturer pricing.
  • Strict quality standards
Interferon gamma-induced protein 10 (IP-10), or CXCL10, is a chemokine secreted by monocytes, endothelial cells and fibroblasts in response to interferon gamma (IFN-ɣ). IP-10 functions as a chemoattractant for activated T cells, monocytes, dendritic, and natural killer (NK) cells that express the G protein-coupled receptor CXCR3. IP-10 is an important factor in autoimmune diseases such as Hashimoto's thyroiditis, Graves' disease, and Type 1 diabetes mellitus. 