Tubing, silicone, versilic
Leverancier: Silex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
686-203 686-206
SILE686-203EA 0 EUR
SILE686-203 SILE686-206
Tubing, silicone, versilic
Slangen Transfer Tubing
Silicone, translucent.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Autoclavable
Used in hospitals and pharmaceutical companies. Ideal for peristaltic pumps.